Learn More
Kimberly-Clark™ Gants en latex
Kimberly-Clark™ KIMTECH PURE™ G5 Latex Glove protects cleanroom users, equipment, and materials from a range of common contaminants.
Marque:  Kimberly-Clark™ HC4411
Code nomenclature Nacres: HA.01
Informations supplémentaires : Poids : 12.50000kg
Code produit. 10007321
  • 240.00€ / 1000 pièces
Expédition estimée
17-03-2020
Ajouter au panier
Description
Description
  • High levels of contamination protection against micro-organisms, chemical splash, particles and extractables
  • Made from non-sterile and silicone-free natural rubber latex
  • Excellent tactile sensitivity and textured finish for handling in delicate processes, in both wet and dry conditions
  • Ambidextrous, powder-free, disposable and featuring a beaded cuff for added strength and easier donning
  • Efficient construction and cleanroom-ready packaging that minimises waste
Spécification
Spécification
Yes
Exam
Yes
Large
Latex Gloves
White
ISO Class 5, ISO16604 , IEST-RP-CC005 EN374-1:2003, EN374-3:2003, AQL 1.5 / EN374-2 Level 2, ISO16604
Beaded/Rolled
Yes
Latex
1000/Pk.
Powdered
30 cm