Learn More
Gibco™ LIF Recombinant Mouse Protein, embryonic stem cell-qualified
Gibco embryonic stem cell (ESC)–qualified mouse LIF is a recombinant protein that supports the culture and maintenance of mouse pluripotent stem cells (PSCs) by suppressing spontaneous differentiation.
Marque: Gibco™ A35933
Informations supplémentaires : Poids : 0.01100kg
Description
Produced in barley grain to minimize endotoxin levels, each lot is carefully tested for purity, microbial contamination, bioactivity, and ability to support mouse PSC culture, thereby minimizing the risk of contamination and helping ensure robust performance.
With LIF Recombinant Mouse Protein, ESC-Qualified, you can:
• Rely on a recombinant growth factor that is qualified for mouse PSC culture
• Safeguard mouse PSC quality with a low-endotoxin LIF
• Culture with confidence with a rigorously tested reagent
Rely on an ESC-qualified growth factor
LIF bioactivity is typically measured by observing its ability to induce differentiation of murine M1 myeloid leukemia cells. In addition to this standard assay, each lot of ESC-qualified mouse LIF is tested for its ability to maintain Oct4 expression in mouse PSCs, thus helping to ensure robust performance for mouse PSC culture.
Safeguard mouse PSC quality with a low-endotoxin LIF
Unlike bacteria-derived recombinant proteins which may carry over harmful bacterial endotoxins, ESC-qualified mouse LIF is derived from plant tissue. It is verified to contain less than 0.005 ng/μg of endotoxin, more than ten times lower than most bacteria-derived counterparts. This unique, low-endotoxin LIF supports the optimal conditions for safeguarding the health and quality of your mouse PSC culture.
Culture with confidence with a rigorously tested reagent
ESC-qualified mouse LIF is tested for bacteria and mycoplasma to minimize the risk of contamination. It is analyzed by SDS-PAGE to ensure >95% purity. It is also tested for pyrogenic and pro-inflammatory contaminants via monocyte activation test.
Spécification
Cell Culture | |
LIF | |
Recombinant | |
Store in freezer (-5°C to -30°C). | |
Room Temperature | |
Cytokines & Receptors | |
Lyophilized | |
>95% by SDS-PAGE |
10 μg | |
Mouse | |
His Tag | |
ES Cell-Qualified | |
Hordeum vulgare (Barley Grain) | |
Recombinant | |
Stem Cell Research |
For Research Use Only. Not for use in diagnostic procedures.