Learn More
Abeba™ Light 1036 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Marque:  Abeba™ 1036/41
Code nomenclature Nacres: HA.03
Informations supplémentaires : Poids : 1.00000kg
Code produit. 15656757
  • 74.80€ / 1 paire
Expédition estimée
17-10-2019
Ajouter au panier
Description
Description
  • CE EN ISO 20345:2011 S1 SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Adjustable instep area
  • Double velcro fastening
  • Suitable for heel and foot lift
Spécification
Spécification
CE EN ISO 20345:2011 S1 SRA
Leather
Slip resistant
1036
Black
41
Polyurethane
Steel Toe