Learn More
Abeba™ Light 1037 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Marque:  Abeba™ 1037/47
Code nomenclature Nacres: HA.03
Informations supplémentaires : Poids : 1.00000kg
Code produit. 15656777
  • 75.00€ / 1 paire
Expédition estimée
18-09-2019
Ajouter au panier
Description
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
Spécification
Spécification
CE EN ISO 20345:2011 S2 SRA
Leather
Slip resistant
1037
Black
47
Polyurethane
Steel Toe