Learn More
Abeba™ Light 31058 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marque:  Abeba™ 31058/42
Code nomenclature Nacres: HA.03
Informations supplémentaires : Poids : 1.11900kg
Code produit. 15658467
  • 83.40€ / 1 paire
Expédition estimée
17-10-2019
Ajouter au panier
Description
Description
  • CE EN ISO 20345:2011 S1 SRA
  • Leather with Air-Mesh textile inlays
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Spécification
Spécification
CE EN ISO 20345:2011 S1 SRA
Leather
Slip resistant
31058
White
42
Polyurethane
Steel Toe