missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lin28A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16177-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is a rabbit polyclonal antibody raised against a peptide mapping within human LIN28.This antibody recognize Lin28A specifically.
Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.Spécification
Lin28A | |
Polyclonal | |
Unconjugated | |
LIN28A | |
CSDD1, LIN 28, lin 28 homolog (C. elegans), Lin 28A, LIN28, Lin28A, Protein lin 28 homolog A, ZCCHC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
500562, 79727, 83557 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K3Y3, Q9H9Z2 | |
Lin28a | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |