Learn More
Lonza™ BioWhittaker™ Hybridoma Serum-Free and Protein-Free Culture Media: UltraDOMA-PF
Click to view available options
Quantité:
500 ml
Conditionnement:
1 pièce
Description
- UltraDOMA-PF is a completely defined medium and does not contain peptides or tissue extracts
1Rehydrates to quantity
Spécification
Spécification
| Quantité | 500 ml |
| Contenu et stockage | 2° to 8°C |