missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRCH1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25017-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein with a leucine-rich repeat and a calponin homology domain. Polymorphism in this gene may be associated with susceptibility to knee osteoarthritis. Alternative splicing results in multiple transcript variants encoding different isoforms.Spécification
LRCH1 | |
Polyclonal | |
Unconjugated | |
LRCH1 | |
CHDC1, KIAA1016, LRCH1, Neuronal protein 81 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23143 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y2L9 | |
LRCH1 | |
LRCH1 Fusion Protein Ag18728 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |