missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC59 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27208-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.Spécification
LRRC59 | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q922Q8, Q96AG4 | |
LRRC59, PRO1855 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
55379, 98238 |
Western Blot | |
Unconjugated | |
LRRC59 | |
Liquid | |
LRRC59 | |
LRRC59 Fusion Protein Ag26090 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human |