missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15593-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes an intracellular F-actin binding protein. The protein is expressed in lymphocytes, neutrophils, macrophages, and endothelium and may regulate neutrophil motility, adhesion to fibrinogen matrix proteins, and transendothelial migration. Alternative splicing results in multiple transcript variants encoding different isoforms.Spécification
LSP1 | |
Polyclonal | |
Unconjugated | |
LSP1 | |
47 kDa actin binding protein, 52 kDa phosphoprotein, LSP1, lymphocyte protein 1, Lymphocyte specific protein 1, pp52, WP34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4046 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P33241 | |
LSP1 | |
LSP1 Fusion Protein Ag7944 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |