Learn More
LXN Mouse anti-Human, Unconjugated, Clone: 3E5A5, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 66957-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829285
  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
Description
The LXN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LXN. This antibody recognizes human, pig, rabbit antigen. The LXN antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
LXN
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig, Rabbit
Western Blot, Immunohistochemistry, ELISA
3E5A5
Unconjugated
20 μL
56925
IgG1