missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G Monoclonal Antibody (1A8-Ly6g), eFluor™ 450, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

121.00€ - 273.00€

Spécification

Antigène Ly-6G
Clone 1A8-Ly6g
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16370934
Afficher les documents Détails de la promotion
Affymetrix eBioscience
48-9668-80
25 μg
121.00€
25µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 16-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16380934
Afficher les documents Détails de la promotion
Affymetrix eBioscience
48-9668-82
100 μg
273.00€
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
En stock
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Description: The monoclonal antibody 1A8-Ly6g reacts with mouse Ly-6G, which is also known as Gr-1. Ly-6G is a 25-kDa GPI protein expressed exclusively by neutrophils. Studies suggest an important role in neutrophil infiltration, recruitment and migration. Ly-6G associates with B2 integrins CD11a and CD11b and can attenuate B2 integrin expression and function. The ligand for Ly-6G is currently unknown. Unlike the RB6-8C5 antibody, 1A8-Ly6g is reported to only bind to Ly-6G found on neutrophils. Moreover, 1A8-Ly6g has been used to deplete neutrophils and reported to have antagonist activity. Applications Reported: This 1A8-Ly6g antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1A8-Ly6g antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.

Ly-6G Monoclonal antibody specifically detects Ly-6G in Mouse samples. It is validated for Flow Cytometry.
Spécification

Spécification

Ly-6G
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
Gr1; Gr-1; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6G
Ly6g
Primary
4° C, store in dark, DO NOT FREEZE!
Ly6g
1A8-Ly6g
Flow Cytometry
eFluor 450
Rat
Mouse
P35461
546644
IgG2a κ
Affinity chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications

For Research Use Only.