missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPLA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16055-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine in both monomeric and micellar forms. The use of alternate polyadenylation sites has been found for this gene. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.Spécification
LYPLA1 | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75608, P70470, P97823 | |
Acyl protein thioesterase 1, APT 1, APT1, hAPT1, LPL I, LPL1, LYPLA1, Lysophospholipase 1, lysophospholipase I, LYSOPLA, LysoPLA I | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
10434, 18777, 25514 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
LYPLA1 | |
Liquid | |
Lypla1 | |
LYPLA1 Fusion Protein Ag9022 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |