Learn More
Thermo Scientific™ M-Series LifterSlips™
Solve the problem of hybridization nonuniformity with Thermo Scientific™ mSeries LifterSlips™. Precise ink printing provides innovative raised edge design for consistent dispersal and easy injection of hybridization solution under the prepositioned LifterSlip. Increased stiffness reduces variation in hybridization fluid volume
Marque:  Thermo Scientific™ KM25IX60I-M-5450
Code nomenclature Nacres: NB.16
Informations supplémentaires : Poids : 0.40000kg
Code produit. 15117246
Cet article n'est plus commercialisé et n'est donc pas disponible. Consultez le produit pour voir les alternatives possibles ou contactez notre support technique au 03 88 67 14 14.
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Specifically drawn for flatness
  • Superior surface quality
Spécification
Spécification
Soda-lime glass
60mm
1mm
35µl
25mm
25 x 60mm
0.75mm
50 pieces in plastic box
Vidéos