missing translation for 'onlineSavingsMsg'
Learn More

Sulfate de magnésium, solution aqueuse 1 M, stérile, Thermo Scientific Chemicals

Catalog Number p-7042008
Click to view available options
Quantité:
125 mL
250 mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
7487-88-9
MgO4S
120.36
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
magnesium(2+) sulfate
[Mg++].[O-]S([O-])(=O)=O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
7487-88-9
MgO4S
120.36
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
magnesium(2+) sulfate
[Mg++].[O-]S([O-])(=O)=O

Magnesium sulfate is applied in replacement therapy for hypomagnesemia. It can be used orally as a laxative to relieve occasional constipation, and to treat low levels of magnesium. It works by replacing magnesium in the body and increasing water in the intestines. It is the first-line antiarrhythmic agent for torsades de pointes in cardiac arrest under the 2005 ECC guidelines and for managing quinidine-induced arrhythmias. It is effective in decreasing the risk that pre-eclampsia progresses to eclampsia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubilité
Entièrement miscible dans l’eau. Soluble dans l’alcool, l’éther (11,6 mg/ml à 18°C) et la glycérine. Insoluble dans l’acétone.

Remarques
Stable dans les conditions de stockage recommandées. Stocker dans des conditions fraîches et sèches et dans des récipients bien scellés.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Magnesium sulfate
Note relative au nom sterile
Forme physique Liquide
Quantité 125 mL
Formule linéaire MgSO4
Informations sur la solubilité Fully miscible in water. Soluble in alcohol,ether (11.6mg/ml at 18°C) and glycerin. Insoluble in acetone.
Poids de la formule 120.37
Concentration or Composition (by Analyte or Components) 1M aq. soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'