missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malectin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26655-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Carbohydrate-binding protein with a strong ligand preference for Glc2-N-glycan. May play a role in the early steps of protein N-glycosylation.Spécification
Malectin | |
Polyclonal | |
Unconjugated | |
MLEC | |
KIAA0152, malectin, MLEC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109154, 304543, 9761 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14165, Q5FVQ4, Q6ZQI3 | |
MLEC | |
Malectin Fusion Protein Ag24379 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |