missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARVELD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13515-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes.Spécification
MARVELD2 | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UZP0, Q8N4S9 | |
DFNB49, MARVD2, MARVEL domain containing 2, MARVELD2, MRVLDC2, Tric, Tricellulin | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
153562, 218518, 365657 |
Western Blot | |
Unconjugated | |
MARVELD2 | |
Liquid | |
Marveld2 | |
MARVELD2 Fusion Protein Ag4446 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human, Rat |