missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCEE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16499-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.Spécification
MCEE | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96PE7, Q9D1I5 | |
DL methylmalonyl CoA racemase, GLOD2, MCEE, methylmalonyl CoA epimerase | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
293829, 73724, 84693 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
MCEE | |
Liquid | |
Mcee | |
MCEE Fusion Protein Ag9665 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |