missing translation for 'onlineSavingsMsg'
Learn More

MCL1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15825-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16895184

  • 129.00€ / 20µL
Expédition estimée: 16-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to MCL1L isoform.

YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
Spécification

Spécification

MCL1L
Polyclonal
Unconjugated
MCL1
EAT, Mcl 1, Mcl 1l, Mcl 1s Delta TM, MCL1, mcl1/EAT, MCL1L, MCL1S, TM
Rabbit
Antigen Affinity Chromatography
RUO
17210, 4170, 60430
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97287, Q07820, Q9Z1P3
Mcl1
Peptide
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions