missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mecr Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51025-2-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The mitochondrial trans-2-enoyl-CoA reductase (MECR), was initially identified as nuclear receptor-binding factor 1 (NRBF1), which can interact with a multitude of nuclear hormone receptors in the presence of the respective ligands. MECR has been shown to be part of the mitochondrial fatty acid synthesis (FAS II) system and to catalyze the NAPDH-dependent reduction of 2-enoyl thioesters, generating saturated acyl-groups. Overexpression of this gene in transgenic mice can lead to cardiac abnormalities, suggesting that inappropriate expression of genes of FAS II can result in the development of hereditary cardiomyopathy.Spécification
Mecr | |
Polyclonal | |
Unconjugated | |
MECR | |
AI195831, Mecr, Nrbf1 | |
Rabbit | |
Protein A | |
RUO | |
26922, 51102 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BV79, Q9DCS3 | |
Mecr | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |