missing translation for 'onlineSavingsMsg'
Learn More

MED12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 20028-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829514

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to MED12.

The initiation of transcription is controlled in part by a large protein assembly known as the preinitiation complex. A component of this preinitiation complex is a 1.2 MDa protein aggregate called Mediator. This Mediator component binds with a CDK8 subcomplex which contains the protein encoded by this gene, mediator complex subunit 12 (MED12), along with MED13, CDK8 kinase, and cyclin C.
Spécification

Spécification

MED12
Polyclonal
Unconjugated
MED12
ARC240, CAG repeat protein 45, CAGH45, FGS1, HOPA, KIAA0192, MED12, mediator complex subunit 12, OKS, OPA containing protein, OPA1, TNRC11, TRAP230
Rabbit
Antigen Affinity Chromatography
RUO
59024, 679693, 9968
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A2AGH6, Q93074
Med12
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions