Learn More
2-aminothiophène-3-carboxylate de méthyle, 97 %, Thermo Scientific Chemicals
Catalog Number p-7036978
Click to view available options
Quantité:
1 g
5 g
4651-81-4
C6H7NO2S
157.19
MFCD00159547
DGGJQLCAYQCPDD-UHFFFAOYSA-N
methyl 2-amino-3-thenoate, methyl 2-amino-3-thiophenecarboxylate, 2-amino-3-methoxycarbonylthiophene, methyl2-aminothiophene-3-carboxylate, 2-amino-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 2-amino-, methyl ester, zlchem 402, pubchem5551, maybridge1_003714, 2-amino-thiophene-3-carboxylicacidmethylester
78381
2-aminothiophène-3-carboxylate de méthyl
COC(=O)C1=C(N)SC=C1
4651-81-4
C6H7NO2S
157.19
MFCD00159547
DGGJQLCAYQCPDD-UHFFFAOYSA-N
methyl 2-amino-3-thenoate, methyl 2-amino-3-thiophenecarboxylate, 2-amino-3-methoxycarbonylthiophene, methyl2-aminothiophene-3-carboxylate, 2-amino-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 2-amino-, methyl ester, zlchem 402, pubchem5551, maybridge1_003714, 2-amino-thiophene-3-carboxylicacidmethylester
78381
2-aminothiophène-3-carboxylate de méthyl
COC(=O)C1=C(N)SC=C1
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Il s’agit d’une importante matière première et d’un intermédiaire utilisé dans la synthèse organique, les produits pharmaceutiques, les produits agrochimiques et les colorants.
Remarques
Conserver à température ambiante. Évitez les agents oxydants forts.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Methyl 2-aminothiophene-3-carboxylate
Point de fusion 76°C to 81°C
Quantité 1 g
Poids de la formule 157.19
Pourcentage de pureté 97%