missing translation for 'onlineSavingsMsg'
Learn More

Méthyle 2-nitro-3,4,5-triméthoxybenzoate, 98 %, Thermo Scientific Chemicals

Catalog Number p-7044396
Click to view available options
Quantité:
10 g
50 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
5081-42-5
C11H13NO7
271.225
MFCD00007097
HCEIEWYPDCDNNU-UHFFFAOYSA-N
methyl 2-nitro-3,4,5-trimethoxybenzoate, methyl 6-nitro-3,4,5-trimethoxybenzoate, 3,4,5-trimethoxy-2-nitrobenzoic acid methyl ester, benzoic acid, 3,4,5-trimethoxy-2-nitro-, methyl ester, 2-nitro-3,4,5-trimethoxybenzoic acid methyl ester, pubchem22545, opera_id_1985, acmc-1ar8w, hceiewypdcdnnu-uhfffaoysa, methyl 3,5-trimethoxy-2-nitrobenzoate
78765
méthyl 3,4,5-triméthoxy-2-nitrobenzoate
COC1=C(C(=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC)OC
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
5081-42-5
C11H13NO7
271.225
MFCD00007097
HCEIEWYPDCDNNU-UHFFFAOYSA-N
methyl 2-nitro-3,4,5-trimethoxybenzoate, methyl 6-nitro-3,4,5-trimethoxybenzoate, 3,4,5-trimethoxy-2-nitrobenzoic acid methyl ester, benzoic acid, 3,4,5-trimethoxy-2-nitro-, methyl ester, 2-nitro-3,4,5-trimethoxybenzoic acid methyl ester, pubchem22545, opera_id_1985, acmc-1ar8w, hceiewypdcdnnu-uhfffaoysa, methyl 3,5-trimethoxy-2-nitrobenzoate
78765
méthyl 3,4,5-triméthoxy-2-nitrobenzoate
COC1=C(C(=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Methyl 2-nitro-3,4,5-trimethoxybenzoate
Point de fusion 66°C to 70°C
Quantité 10 g
Beilstein 2702653
Poids de la formule 271.23
Pourcentage de pureté 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'