Learn More
Thermo Scientific Chemicals Vert de méthyle, sel de chlorure de zinc, pur, certifié
Catalog Number p-4229039
Click to view available options
Quantité:
25 g
100 g
Conditionnement:
Flacon en verre
7114-03-6
C27H35BrClN3·xZnCl2
653.23
7114-03-6
C27H35BrClN3·xZnCl2
653.23
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Methyl Green, zinc chloride salt
Point de fusion 233.0°C
Couleur Marron-rouge ou vert
Forme physique Poudre cristalline
Quantité 25 g
Merck Index 14,6082
Synonyme C.I. 42590, Ethyl Green, zinc chloride salt
Informations sur la solubilité Solubility in water: 70g/L (20°C)
Clé InChI ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
Nom IUPAC zinc ;[4-[[4-(diméthylamino)phényl]-(4-diméthylazaniumylidénécyclohexa-2,5-dien-1-ylidène)méthyl]phényl]-éthyl-diméthylazanium ;bromure ;trichlorure
CID PubChem 131675247
Poids de la formule 653.23
Qualité Pures