missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Marque: ROBU VitraPOR™ 18000
Informations supplémentaires : Poids : 0.02140kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Spécification
Borosilicate Glass 3.3 | |
6 mm | |
15 mm |
0 | |
Non-sterile |