missing translation for 'onlineSavingsMsg'
Learn More

Mitofilin Monoclonal Antibody (MM#7A-2E4AD5), Alexa Fluor™ 488, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen MA456400A488

Informations supplémentaires : Poids : 0.01000kg

Code produit. 15984334

  • 471.00€ / 50µL
Expédition estimée: 16-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Mitofilin Monoclonal antibody specifically detects Mitofilin in Human samples. It is validated for Immunocytochemistry.
Spécification

Spécification

Mitofilin
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Q16891
IMMT
Human heart mitochondria
50 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry
MM#7A-2E4AD5
Alexa Fluor 488
IMMT
1700082C19Rik; cell proliferation-inducing gene 4/52 protein; cell proliferation-inducing protein 52; D830041H16Rik; EMBL:AAI05842.1, ECO:0000312; EMBL:AAI05842.1}; HMP; Immt; immt {ECO:0000312; IMMT antibody; inner membrane mitochondrial protein; inner membrane protein, mitochondrial; inner membrane protein, mitochondrial (mitofilin); MGC133830 protein; MIC60; MICOS complex subunit Mic60; MINOS2; mitochondrial inner membrane organizing system 2; mitochondrial inner membrane protein; mitochondrial inner membrane protein {ECO:0000250; Mitofilin; mitofilin {ECO:0000250; motor protein; P87; p87/89; p87/89 antibody; P89; PIG4; PIG52; proliferation-inducing gene 4; RGD:1310684}; UniProtKB:Q8CAQ8, ECO:0000312; UniProtKB:Q8CAQ8}
Mouse
purified
RUO
10989
4° C, store in dark, DO NOT FREEZE!
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions