missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKK7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55030-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Coronins, such as CORO7, constitute an evolutionarily conserved family of WD-repeat actin-binding proteins. CORO7 plays a role in Golgi complex morphology and function.Spécification
MKK7 | |
Polyclonal | |
Unconjugated | |
MAP2K7 | |
JNK activating kinase 2, JNK kinase 2, JNKK 2, Jnkk2, MAP kinase kinase 7, MAP2K7, MAPK/ERK kinase 7, MAPKK 7, MAPKK7, MEK 7, MEK7, MKK7, PRKMK7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26400, 5609 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14733, Q8CE90 | |
Map2k7 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |