missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16206-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene localizes to the basal body and is required for formation of the primary cilium in ciliated epithelial cells. Mutations in this gene result in Meckel syndrome type 1 and in Bardet-Biedl syndrome type 13. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
MKS1 | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q499Q5, Q5SW45, Q9NXB0 | |
BBS13, Meckel syndrome type 1 protein, Meckel syndrome, type 1, MES, MKS, MKS1 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
287612, 380718, 54903 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
MKS1 | |
Liquid | |
Mks1 | |
MKS1 Fusion Protein Ag9177 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |