missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13975-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
MKS3 | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5HYA8, Q8BR76 | |
JBTS6, Meckel syndrome type 3 protein, MECKELIN, MKS3, TMEM67, TNEM67, transmembrane protein 67 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
329795, 91147 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
TMEM67 | |
Liquid | |
Tmem67 | |
MKS3 Fusion Protein Ag5009 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |