missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24395-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
CPSF1 is a component of the cleavage and polyadenylation specificity factor (CPSF) complex that plays a key role in pre-mRNA 3′-end formation, recognizing the AAUAAA signal sequence and interacting with poly(A) polymerase and other factors to bring about cleavage and poly(A) addition. This subunit is involved in the RNA recognition step of the polyadenylation reaction.Spécification
MLK4 | |
Polyclonal | |
Unconjugated | |
MAP3K21 | |
KIAA1804, mixed lineage kinase 4, MLK4, RP5 862P8.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234878, 307950, 84451 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5TCX8, Q8VDG6 | |
MAP3K21 | |
MLK4 Fusion Protein Ag18904 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |