Learn More
MLX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA562677
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: EEVSTLRKDV TALKIMKVNY EQIVKAHQDN PHEGEDQVSD QVKFNVFQGI MDSLFQSFNA SISVASFQEL SA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Spécification
MLX | |
Polyclonal | |
Unconjugated | |
Mlx | |
bHLHd13; BigMax alpha; BigMax protein; class D basic helix-loop-helix protein 13; hypothetical protein LOC508531; im:7146242; MAD7; MAX dimerization protein MLX; MAX-like bHLHZIP protein; max-like factor protein; max-like protein X; Mlx; MLX, MAX dimerization protein; MXD7; Protein BigMax; TCFL4; TF4; transcription factor like 4; transcription factor-like 4; transcription factor-like protein 4; wu:fb61a01; wu:fb99c01; zgc:112409 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UH92 | |
Mlx | |
Recombinant protein corresponding to Human MLX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only