missing translation for 'onlineSavingsMsg'
Learn More

MME/CD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23898-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802515

  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD10 (Common Acute Lymphocytic Leukemia Antigen,CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt′s, and follicular germinal center lymphomas, immature B cells with in adult bone marrow and on cells from patients with chronic myelocytic leukemia (CML). CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells. CD10 is a neutral endopeptidase that cleaves peptides at the amino side of hydrophobic residues and inactivates several peptide hormones including glucagon, enkephalins, substance P, neurotensin, oxytocin, and bradykinin. Further, CD10 is a 100 kDa type II transmembrane glycoprotein that exists in a single copy of greater than 45 kb. The 5′ untranslated region of the CD10 gene is alternatively spliced, resulting in four separate mRNA transcripts and the coding region is not affected by alternative splicing. Diseases associated with CD10 dysfunction include spinocerebellar ataxia 43 and Charcot-Marie tooth Disease.
Spécification

Spécification

MME/CD10
Polyclonal
Unconjugated
MME
Atriopeptidase, CALLA, CD10, Enkephalinase, EPN, membrane metallo endopeptidase, MME, NEP, Neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11
Rabbit
Antigen Affinity Chromatography
RUO
17380, 24590, 4311
-20°C
Liquid
Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07861, P08473, Q61391
Mme
MME,CD10 Fusion Protein Ag20963
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions