missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOBKL2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16942-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
MOBKL2A | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BSU7, Q96BX8 | |
MOB LAK, Mob1 homolog 2A, MOB3A, MOBKL2A, moblak, Protein Mob3A | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
126308, 208228, 362833 |
Western Blot | |
Unconjugated | |
MOB3A | |
Liquid | |
Mob3a | |
MOBKL2A Fusion Protein Ag10540 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |