Learn More
Invitrogen™ Human Monoamine Oxidase B, Recombinant Protein
Recombinant Protein
Marque: Invitrogen™ RP75659
Code nomenclature Nacres: NA.28
Informations supplémentaires : CAS : 69-74-9 Poids : 0.01000kg
Description
Monoamine Oxidase B (human) Recombinant Protein, RP-75659, from Invitrogen.
This product contains purified recombinant human Monoamine Oxidase B protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽59.84 kDa.Spécification
P27338 | |
Control, Western Blot | |
4129 | |
8 | |
E. Coli | |
RUO | |
Human Monoamine Oxidase B | |
Human | |
Hu | |
Recombinant | |
Recombinant |
0.268 mg/mL | |
TBS with .01mM EDTA, 50% glycerol and no preservative; pH 8 | |
Human Monoamine Oxidase B | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
MAO b | |
MAOB | |
Unconjugated | |
E. coli | |
Liquid | |
purified |