missing translation for 'onlineSavingsMsg'
Learn More

MORF4L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 55257-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836755

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody has no cross-reaction to MORF4L2.

Cellular senescence is the terminal non-dividing state that normal cells enter following completion of their proliferative potential. Fusions of immortal human cell lines with each other have led to their assignment to one of four complementation groups. Mortality factor 4 (MORF4) was identified as the lead member of a family of transcription factor-like proteins that reverses this immortal phenotype. Like other members in this family, MORF4 is localized to the nucleus and possesses transcription factor-like motifs such as helix-loop-helix and a leucine zipper motif that might allow it to form transcriptionally active homo- or heterodimers. MORF4 has been shown to bind to the transcription corepressors mSin3A and TLE, suggesting that together, these complexes may play a role in transcriptional repression of genes that lead to cellular senescence.
Spécification

Spécification

MORF4L1
Polyclonal
Unconjugated
MORF4L1
Eaf3, FWP006, HSPC008, HSPC061, HsT17725, MORF related gene 15 protein, MORF4L1, MORFRG15, mortality factor 4 like 1, MRG15, PP368, Protein MSL3 1, S863 6
Rabbit
Antigen Affinity Chromatography
RUO
10933, 21761, 300891
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.55 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P60762, Q6AYU1, Q9UBU8
Morf4l1
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions