Learn More
ACROBiosystems Mouse CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marque: ACROBiosystems CD7-M5252-100UG
Informations supplémentaires : Poids : 0.00001kg
Description
CD Antigen CD27 is also known as Tumor necrosis factor receptor superfamily member 7 (TNFRSF7), which belongs to TNF-receptor superfamily. CD27 / TNFRSF7 is found in most T-lymphocytes. CD27 / TNFRSF7 is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. This receptor transduces signals that lead to the activation of NF-κB and MAPK8/JNK. CD27 has been shown to interact with SIVA1, TRAF2 and TRAF3.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001028298.1 | |
100 μg | |
Thr 21 - Arg 182 | |
CD27,TNFRSF7,S152,T14,Tp55 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
45.1 kDa | |
Mouse CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-M5252) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Arg 182 (Accession # P41272-1). | |
-20°C | |
CD27 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |