missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse CLEC12A / MICL / CLL-1 Protein, His Tag
423.38€ - 3108.82€
Spécification
Poids moléculaire | 25.7 kDa |
---|---|
Source | Mouse CLEC12A, His Tag (CLA-M5244) is expressed from human 293 cells (HEK293). It contains AA Tyr 65 - Arg 267 (Accession # Q504P2-1). |
Immunogène | Tyr 65 - Arg 267 |
Conditions de stockage | -20°C |
Alias de gène | CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17256180
|
ACROBiosystems
CLA-M5244-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17266180
|
ACROBiosystems
CLA-M5244-1MG |
1 mg |
3108.82€
1mg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CLEC12A (C-type lectin domain family 12 member A) is also known as CLL1, DCAL2, MICL. Clec12a is an inhibitory receptor for uric acid crystals that regulates inflammation in response to cell death. Cell surface receptor that modulates signaling cascades and mediates tyrosine phosphorylation of target MAP kinases. Evidence of distinct disease propagating stem cells in myelodysplastic syndrome (MDS) has emerged in recent years. The role of CLEC12A in MDS, however, remains to be elucidated. Furthermore, CLEC12A has been proposed as a promising marker of leukaemic stem cells in AML.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.7 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
25.7 kDa | |
Tyr 65 - Arg 267 | |
CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
Mouse CLEC12A, His Tag (CLA-M5244) is expressed from human 293 cells (HEK293). It contains AA Tyr 65 - Arg 267 (Accession # Q504P2-1). | |
-20°C | |
CLEC12A | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers & CD Antigens |