missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse gp130 / CD130 / IL-6 R beta Protein, His Tag

Marque:  ACROBiosystems ILT-M52H1-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15867137

  • 1815.83€ / 1mg
Date d’expédition estimée: 07-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Interleukin-6 receptor subunit beta (IL6ST) is also known as IL-6 receptor subunit beta, IL-6R subunit beta, IL-6R-beta, IL-6RB, Interleukin-6 signal transducer, Membrane glycoprotein 130 (gp130), CD130, Oncostatin-M receptor subunit alpha and Il6st,which is single-pass type I membrane protein. IL6ST /gp130 /CD130 can be found in tissues such as brain, heart, thymus, spleen, kidney, lung and liver and found in all the cell lines tested except BaF-B03.The expression of IL-6ST /gp130 is not restricted to IL6-responsive cells. The receptor systems for IL6, LIF, OSM, CNTF, IL11, CTF1 and BSF3 can utilize gp130 for initiating signal transmission. IL6ST /CD130 can bind to IL6 /IL6R (alpha chain) complex, resulting in the formation of high-affinity IL6 binding sites, and transduce the signal. IL6ST /GP130 does not bind IL6 and may have a role in embryonic development.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 68.5 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_034690
HEK293
Mouse
68.5 kDa
1 mg
Gln 23 - Glu 617
IL6ST,gp130,CD130,IL-6 RB,IL-6 R-beta,CDw130
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Glu 617 (Accession # NP_034690).
-20°C
gp130
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions