missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse IgG1 Fc Protein, Tag Free
326.60€ - 846.76€
Spécification
Numéro d’adhésion | AAK53870.1 |
---|---|
Poids moléculaire | 24.3 kDa |
Source | Mouse IgG Fc, Tag Free (IG1-M5208) is expressed from human 293 cells (HEK293). It contains AA Val 111 - Lys 324 (Accession # AAK53870.1). |
Immunogène | Val 111 - Lys 324 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876987
|
ACROBiosystems
IG1-M5208-200UG |
200 μg |
326.60€
200µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15866987
|
ACROBiosystems
IG1-M5208-1MG |
1 mg |
846.76€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.
This protein carries no "e;tag"e;. The protein has a calculated MW of 24.3 kDa. The protein migrates as 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAK53870.1 | |
Mouse IgG Fc, Tag Free (IG1-M5208) is expressed from human 293 cells (HEK293). It contains AA Val 111 - Lys 324 (Accession # AAK53870.1). | |
-20°C | |
IgG1 Fc | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
24.3 kDa | |
Val 111 - Lys 324 | |
IgG1 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |