missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse IgG1 Fc Protein, Tag Free

Marque:  ACROBiosystems IG1-M5208-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15866987

  • 605.28€ / 1mg
Date d’expédition estimée: 15-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.

This protein carries no "e;tag"e;. The protein has a calculated MW of 24.3 kDa. The protein migrates as 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAK53870.1
HEK293
Mouse
24.3 kDa
1 mg
Val 111 - Lys 324
IgG1
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Mouse IgG Fc, Tag Free (IG1-M5208) is expressed from human 293 cells (HEK293). It contains AA Val 111 - Lys 324 (Accession # AAK53870.1).
-20°C
IgG1 Fc
Tag Free
Vidéos
FDS
Documents
Certificats
Promotions

Promotions