missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse IgG2a Fc Protein, Tag Free
326.60€ - 846.76€
Spécification
Numéro d’adhésion | CAC20702 |
---|---|
Poids moléculaire | 26.4 kDa |
Source | Mouse IgG2a Fc, Tag Free (IGA-M5207) is expressed from human 293 cells (HEK293). It contains AA Glu 98 - Lys 330 (Accession # CAC20702). |
Immunogène | Glu 98 - Lys 330 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867007
|
ACROBiosystems
IGA-M5207-1MG |
1 mg |
846.76€
1mg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15877007
|
ACROBiosystems
IGA-M5207-200UG |
200 μg |
326.60€
200µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Immunoglobulin G2 (IgG2) is a member of many immunoglobulin G developed and secreted by effective B cells. In wake of cutting by pepsin, IgG is divided into two F(ab)s with one antigen binding site and a high conserved Fc segment. The Fc segment bears a highly conserved N-glycosylation site. There are two members of IgG2: IgG2a and IgG2b. It was found that IgG2a was superior to IgG1 in activating complement. The glycosylation of the circulating immunoglobulin-γ (IgG) antibody molecules changes in rheumatoid arthritis.
This protein carries no "e;tag"e;. The protein has a calculated MW of 26.4 kDa. As a result of glycosylation, the protein migrates as 30-35 kDa under reducing (R) condition, and 55-60 kDa under non-reducing (NR) condition (SDS-PAGE).Spécification
CAC20702 | |
Mouse IgG2a Fc, Tag Free (IGA-M5207) is expressed from human 293 cells (HEK293). It contains AA Glu 98 - Lys 330 (Accession # CAC20702). | |
-20°C | |
IgG2a Fc | |
Mouse | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
26.4 kDa | |
Glu 98 - Lys 330 | |
IgG2A | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |