missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-17F Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ 210-17F-250UG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17890423

  • 1010.00€ / 250µg
Expédition estimée: 25-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

IL-17F (Interleukin 17F, CTLA-8)) is a cytokine belonging to the IL-17 family that is produced by inflammatory cells such as activated T cells, mast cells, and basophils. IL-17F is involved in allergic airway inflammation, and can induce several cytokines, chemokines, and adhesion molecules in bronchial epithelial cells, vein endothelial cells, fibroblasts, and eosinophils. IL-17F may be secreted as a homodimer, or a heterodimer with IL17A. It acts by binding to the type I receptor, IL-17R, aiding recruitment of monocytes and neutrophils at the site of inflammation by increasing chemokine production. IL-17F also stimulates induction of other pro-inflammatory cytokines TNF alpha, IL-1 beta, IL-6, and IL-8, and reports strongly suggest the involvement of IL-17 in several chronic inflammatory diseases such as rheumatoid arthritis, psoriasis and multiple sclerosis. TGF-beta (differentiation) and IL-23 (expansion) are required for induction and maintenance of Th17 (IL-17 producing) cells, which in turn induce the other pro-inflammatory cytokines. IL-17F is produced, and exists, as a homo-dimer, with homology to a herpes virus early protein, is one of the six members (IL-17A-F) of this cytokine family, and is well characterized and highly expressed by activated effector memory T cells. IL-17F has been found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1.
Spécification

Spécification

Q7TNI7
protein with no preservative
30
250 µg
-20°C
<1 EU/µg
IL17F
Unconjugated
MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
≥ 98% by SDS-PAGE gel and HPLC analyses.
Functional Assay
257630
Mouse IL-17 F
E. coli
RUO
C87042; CANDF6; cytokine ML-1; IL17F; IL-17 F; IL-24; ILN; Interleukin; interleukin 17 F; Interleukin17F; interleukin-17 F; interleukin-24; ML1; ML-1; mutant IL-17 F
Mouse
Recombinant
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions