missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse LAG-3 / CD223 Protein, His Tag

Marque:  ACROBiosystems LA3-M52H5-100UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877217

  • 353.08€ / 100µg
Date d’expédition estimée: 28-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.1 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by SDS-PAGE
Mouse LAG-3, His Tag (LA3-M52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession # Q61790).
-20°C
LAG-3
His Tag
HEK293
Mouse
47.1 kDa
100 µg
Gly 24 - Leu 442
LAG3,CD223,FDC
Unconjugated
PBS, pH7.4
Vidéos
FDS
Documents
Certificats
Promotions

Promotions