Learn More
MP Biomedicals™ Collagen from Calf Skin
Soluble form
Marque:  MP Biomedicals™ 0216008483
Code nomenclature Nacres: NA.28
Informations supplémentaires : CAS : 9007-34-5 Poids : 0.00030kg
Code produit. 11490272
  • 546.96€ / 250mg
Expédition estimée: 03-06-2024
Description
Suitable for gel formation, platelet aggregation and assay of collagenase by viscometry. Prepared by method of Gallop, P.M. and Seifter, S., Meth. in Enzymol., VI: 635 (1963).
Spécification
Spécification
5mg/mL
∽1000000
3.7
Calf Skin
Solid
Gel Formation, Platelet Aggregation, Collagenase Assay
Collagen from Calf skin
250mg
4°C