Learn More
MP Biomedicals™ Collagen from Calf Skin
Soluble form
Marque: MP Biomedicals™ 0216008483
Code nomenclature Nacres: NA.28
Informations supplémentaires : CAS : 9007-34-5 Poids : 0.00030kg
Description
Suitable for gel formation, platelet aggregation and assay of collagenase by viscometry. Prepared by method of Gallop, P.M. and Seifter, S., Meth. in Enzymol., VI: 635 (1963).
Spécification
5mg/mL | |
∽1000000 | |
3.7 | |
Calf Skin | |
Solid |
Gel Formation, Platelet Aggregation, Collagenase Assay | |
Collagen from Calf skin | |
250mg | |
4°C |