missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14715-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
MPP1 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70290, Q00013 | |
AAG12, DXS552, DXS552E, EMP55, MPP1, MRG1, p55, PEMP | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
17524, 4354, 652956 |
Western Blot | |
Unconjugated | |
MPP1 | |
Liquid | |
Mpp1 | |
MPP1 Fusion Protein Ag6439 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human, Rat |