Learn More
MPZL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25513-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This polyclonal antibody raised against 32-235 aa of human MPZL3 reveals bands of 28 kDa and 70 kDa, which could be endogenous MPZL3 of unmodified form and post-translational modified form, respectively.
MPZL3 mediates homophilic cell-cell adhesion.Spécification
MPZL3 | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3V3F6, Q6UWV2 | |
MPZL3, myelin protein zero like 3 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
196264, 319742 |
Western Blot | |
Unconjugated | |
MPZL3 | |
Liquid | |
Mpzl3 | |
MPZL3 Fusion Protein Ag21905 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |