missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL33 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23870-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Alternatively spliced transcript variants encoding different isoforms have been described.Spécification
MRPL33 | |
Polyclonal | |
Unconjugated | |
MRPL33 | |
C2orf1, L33mt, MRP L33, MRPL33, RPL33L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9553 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75394 | |
MRPL33 | |
MRPL33 Fusion Protein Ag20921 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |