missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14973-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a myotubularin-related protein. The encoded protein is a phosphoinositide phosphatase that specifically dephosphorylates phosphatidylinositol 3,5-biphosphate and phosphatidylinositol 3-phosphate. Mutations in this gene are correlated with autosomal domit centronuclear myopathy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 18.Spécification
MTMR14 | |
Polyclonal | |
Unconjugated | |
MTMR14 | |
C3orf29, hEDTP, HJUMPY, MTMR14, NS5ATP4ABP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
312634, 64419, 97287 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Flow Cytometry, Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NCE2, Q8VEL2 | |
Mtmr14 | |
MTMR14 Fusion Protein Ag6883 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |