missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTP18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14257-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
MTP18 (mitochondrial 18 kDa protein), also known as HSPC242, is a 166 amino acid protein essential for cell viability. MTP18 may also have a role in maintaining the integrity of the mitochondrial network. Localized to the mitochonrion, expression of MTP18 is regulated by the PI 3-kinase pathway. Loss of function of MTP18 results in the release of cytochrome c, which subsequently activates the caspase cascade and leads to apoptosis.Spécification
MTP18 | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CRB8, Q9UDX5 | |
HSPC242, Mitochondrial 18 kDa protein, mitochondrial protein 18 kDa, MTP18, My022 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
289745, 51537, 67900 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
MTFP1 | |
Liquid | |
Mtfp1 | |
MTP18 Fusion Protein Ag5530 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |