Learn More
Novus Biologicals™ Multi-Species dsRNA ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marque: Novus Biologicals™ NBP3-11368-200Tests
892.00 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Spécification
1 - 300 ng/ml (0.1 - 30 ng/well) | |
ELISA Kit (Colorimetric) | |
ELISA | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
Multi-Species |
3 ng/ml | |
The dsRNA Detection Kit allows sensitive and selective detection of dsRNA molecules (larger than 30-40 bp), independent of their nucleotide composition and sequence. | |
200 Tests | |
Double-stranded RNA |